This product is now discontinued, View Related Products

Combs for Thermo Scientific Owl Horizontal Large-Format

Use combs to produce the wells you need for your electrophoresis application. Select the comb based on the gel width and the number of wells.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Description
Availability Pricing
Thermo Scientific A1-12D Replacement Comb for Electrophoresis Gels, 13 cm W, 12 Wells; 1/EA EW-28553-11 Mfr # A1-12D
COMPARE
12 Replacement Comb for 28553-00,-10 12 wells 1.5mm gel
Call For Price
Thermo Scientific A1-16D Replacement Comb for Electrophoresis Gels, 13 cm W, 16 Wells; 1/EA EW-28553-12 Mfr # A1-16D
COMPARE
16 Replacement Comb for 28553-00,-10 16 wells 1.5mm gel
Call For Price
Thermo Scientific A1-20D Replacement Comb for Electrophoresis Gels, 13 cm W, 20 Wells; 1/EA EW-28553-13 Mfr # A1-20D
COMPARE
20 Replacement Comb for 28553-00,-10 20 wells 1.5mm gel
Call For Price
Thermo Scientific A2-16D Replacement Comb for Electrophoresis Gels, 20 cm W, 16 Wells; 1/EA EW-28553-31 Mfr # A2-16D
COMPARE
16 Replacement Comb for 28553-20,-30 16 wells 1.5mm gel
Call For Price
Thermo Scientific A2-24D Replacement Comb for Electrophoresis Gels, 20 cm W, 24 Wells; 1/EA EW-28553-32 Mfr # A2-24D
COMPARE
24 Replacement Comb for 28553-20,-30 24 wells 1.5mm gel
Call For Price
Thermo Scientific A2-36D Replacement Comb for Electrophoresis Gels, 20 cm W, 36 Wells; 1/EA EW-28553-33 Mfr # A2-36D
COMPARE
36 Replacement Comb for 36 wells 1.5mm gel
Call For Price
Thermo Scientific D3-MTD Replacement Comb for Electrophoresis Gels, 23 cm W, 25 Wells; 1/EA EW-28558-51 Mfr # D3-MTD
COMPARE
25 Replacement Comb for 28553-40, 25 wells, 1.5mm gel
Call For Price
Thermo Scientific D3-MT2D Replacement Comb for Electrophoresis Gels, 23 cm W, 50 Wells; 1/EA EW-28558-53 Mfr # D3-MT2D
COMPARE
50 Replacement Comb for 28553-40, 50 wells, 1.5mm gel
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat