This product is now discontinued, View Related Products
Compound Microscopes with Infinity-Corrected Optics
Infinity-corrected optics give excellent images with enhanced contrast
This compound microscope features lenses that improve image clarity — ideal for biological applications. Viewing head has 45° inclined eyepieces with diopter and interpupillary adjustment. Siedentopf type head ensures the tube length remains the same when adjusting interpupillary distance. Feature a 20-watt halogen bulbs and an Abbe condenser.
Microscopes feature coarse and fine focusing adjustments, and large 5-1/2" x 5-1/2" (140 x 140 mm) stage with specimen holder. Objective turret is reverse-mounted giving you easy access to the stage.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Head Configuration Maximum Magnification
Availability Pricing
Swift Optical M15B-P Binocular Microscope w/Infinity-Corrected Optics; 40x/100x EW-48924-70 Mfr # M15B-P
COMPARE
Binocular 1000x
Call For Price
Swift Optical M15B-P Trinocular Microscope w/Infinity-Corrected Optics; 400x/1000x EW-48924-71 Mfr # M15T-P
COMPARE
Trinocular 1000x