Compression Straight Unions
Very good chemical resistance. Rates to 220 psi at 77ºF ( 25ºC). Usable from 32 to 212ºF (0 to 100ºC).
ITEM(S) ON SALE
$8.90 - $693.00USD / Pkg of 10
58 variations of this product are available.
See All Product Options Below
58 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
1/4 1/4 Polypropylene
$40.05USD / Pkg of 10
COMPARE
3/8 3/8 PVDF
$14.10USD / Each
COMPARE
3/4 3/4 PVDF
$55.70USD / Each
COMPARE
1/2 1/2 Nylon
$29.40USD / Pkg of 10
$36.75USD / Pkg of 10
COMPARE
5/8 5/8 Nylon
$41.90USD / Pkg of 10
Parker Hannifin Compression fittings, Straight unions; Black PP,1/4" OD, 1-5/8" x 11/16" x 5/32" EW-06444-51 Mfr # P4UC4
COMPARE
1/4 1/4 Polypropylene
$74.72USD / of 25
$93.40USD / of 25
Parker Hannifin Compression fittings, Straight unions; Black PP,3/8" OD, 1-13/16" x 3/16" x 1/4"
Buy More Save More
EW-06444-52 Mfr # P6UC6
COMPARE
3/8 3/8 Polypropylene
$123.00USD / of 25
Parker Hannifin Compression Union, 1/8" x 1/8", Stainless Steel, Each
Buy More Save More
EW-30486-00 Mfr # 2SC2-316
COMPARE
1/8 1/8 316 Stainless Steel
$26.70USD / Each
Parker Hannifin 12SC12-316 Compression Union, 1/16" x 3/4", Stainless Steel, Each
Buy More Save More
EW-30486-06 Mfr # 12SC12-316
COMPARE
3/4 3/4 316 Stainless Steel
$87.50USD / Each
COMPARE
1/4 1/4 Polypropylene
$117.00USD / Pkg of 100
COMPARE
1/4 1/4 Nylon
$296.00USD / Pkg of 100
COMPARE
3/8 3/8 Nylon
$271.00USD / Pkg of 100
COMPARE
1/2 1/2 Nylon
$351.00USD / Pkg of 100
COMPARE
1/2 1/2 PVDF
$83.90USD / Pkg of 10
COMPARE
3/4 3/4 PVDF
$693.00USD / Pkg of 10
COMPARE
1/8 1/8 316 Stainless Steel
$24.00USD / Each
COMPARE
3/8 3/8 316 Stainless Steel
$33.40USD / Each
COMPARE
1/2 1/2 316 Stainless Steel
$45.80USD / Each
COMPARE
3/4 3/4 316 Stainless Steel
$67.30USD / Each
COMPARE
1/2 1/2 Brass
$17.30USD / Each
Showing 1 - 20 of 58