Contact Plate Media
Cultivate microorganisms from environmental surfaces
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
These high-quality, economical plates feature a specified grid molded into the bottom of the plate for easy colony counting.
$27.50 - $51.50USD / Bag of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Media Name (No.)
Availability Pricing
COMPARE
Tryptic Soy w/ Lecithin, Tween
$27.50USD / Bag of 10
COMPARE
SabDex (Sabourand Dextrose) agar
$39.70USD / Bag of 10
COMPARE
Blood
$51.50USD / Bag of 10
COMPARE
Eosin Methylene Blue
$27.50USD / Bag of 10