Contacting Epoxy Body Conductivity Cells
Ideal for laboratory or light industrial use
  • Graphite measuring surfaces
  • PT 1000 RTD temperature compensation
  • 3/4" M NPT mounting colar
Economical laboratory or light industrial conductivity sensors feature either graphite or platinum ring technology. The durable epoxy body construction provides a rugged, dependable sensor. Suitable for most conductivity meters.
$293.00 - $309.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cable Length (ft) Cell Constant
Availability Pricing
Sensorex FC75P Conductivity Probe, K=10 with ATC-P1K, 3/4"NPT(M) EW-19066-18 Mfr # CS200TC-K=10 + FC75P
COMPARE
10 K = 10
$293.00USD / Each
Sensorex FC75P Conductivity Probe, K=0.1 with ATC-P1K, 3/4"NPT(M); 20' Cable EW-19066-26 Mfr # 971972
COMPARE
20 K = 0.1
$309.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat