This product is now discontinued, View Related Products
COORSTEK Buchner Vacuum Funnels, Ceramic
One piece tabletop filter with side outlet for vacuum filtration accepts standard filters. Funnels are glazed inside and out except for the rim. Tubulature OD is approximately 14.5 mm at widest point. Autoclavable.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Filter Paper Diameter (mm) Material
Availability Pricing
CoorsTek 66281 Buchner Vacuum Funnels, Ceramic, 750 mL, 1/pk EW-17817-02 Mfr # 66281
COMPARE
750 124 Ceramic
Call For Price
CoorsTek 60281 Buchner Vacuum Funnels, Ceramic, 750 mL, 2/cs EW-17817-22 Mfr # 66281
COMPARE
750 Ceramic
Call For Price
CoorsTek 66282 Buchner Vacuum Funnels, Ceramic, 1900 mL, 1/cs EW-17817-24 Mfr # 66282
COMPARE
1900 185 Ceramic
Call For Price
CoorsTek 66283 Buchner Vacuum Funnels, Ceramic, 4000 mL, 1/cs EW-17817-26 Mfr # 66283
COMPARE
4000 240 Ceramic