CoorsTek Mortars, Alumina
Ideal for precise elemental analysis where minimal sample contamination is required. Mortars are glazed all over except for grinding surfaces and the outside bottom. Features an octagonal shape. Mortar and pestle are sold as separate pieces. Autoclavable.
$153.00USD / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Mortars Capacity (mL) Mortars Top OD (mm)
Availability Pricing
CoorsTek 60335 Alumina Mortar, 1.5 mL, pack of 1 EW-17822-00 Mfr # 60335
COMPARE
1.5 35
Call For Price
CoorsTek 60358 Alumina Mortar, 7 mL, pack of 1 EW-17822-02 Mfr # 60358
COMPARE
7 50
$153.00USD / Pkg of 1
CoorsTek 60361 Alumina Mortar, 15 mL, pack of 1 EW-17822-04 Mfr # 60361
COMPARE
15 65
Call For Price
CoorsTek 60358 Alumina Mortar, 7 mL, case of 3 EW-17822-22 Mfr # 60358
COMPARE
7 50
Call For Price
CoorsTek 60364 Alumina Mortar, 30 mL, case of 1 EW-17822-26 Mfr # 60364
COMPARE
30 75
Call For Price
CoorsTek 60370 Alumina Mortar, 80 mL, case of 1 EW-17822-28 Mfr # 60370
COMPARE
80 100
Call For Price
CoorsTek 60373 Alumina Mortar, 120 mL, case of 1 EW-17822-30 Mfr # 60373
COMPARE
120 120
Call For Price
Showing 1- 7 of 7