Copper Tubing for Gas Chromatography Lines

Ideal for connecting gas lines in gas chromatographs
  • Chemically compatible with FREON® and noncorrosive chemicals
Applications: Excellent for use with FREON® and noncorrosive chemicals. Ideal for connecting gas lines to gas chromatographs and for general plumbing applications.
Characteristics: Seamless. Soft-tempered design allows you to cut tubing to desired length.
Certification: None.
Temperature range: –60 to 400°F (–53 to 204°C)
Sterilize: With autoclave, ethylene oxide, or gamma irradiation.
$538.50 - $642.50USD / Roll of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
3100 1/16 1/8
$538.50USD / Roll of 1
COMPARE
1450 3/16 1/4
$642.50USD / Roll of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat