Corning 96-Well Black and White Solid Plates
  • Tested on a lot-to-lot basis for opacity and low background fluorescence/luminescence—assures minimal well-to-well crosstalk
  • Choose Black Polystyrene plates for minimizing background fluorescence and backlight scatter.
  • Well volume is 360 µL.
$586.00 - $725.00USD / Case of 100
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Sterile Surface Treatment Well Shape
Availability Pricing
Corning 3789 Plate,round,white,100/cs, (White) EW-01728-60 Mfr # 3789A
COMPARE
Untreated Round
$646.00USD / Case of 100
Corning 3916 Plate,treated,black,100/cs, Black EW-01728-62 Mfr # 3916
COMPARE
Yes Cell culture Flat
$707.00USD / Case of 100
Corning 3917 Plate,treated,white,100/cs, White EW-01728-63 Mfr # 3917
COMPARE
Yes Cell culture Flat
$674.00USD / Case of 100
Corning 3922 96-Well White Solid Plates, High binding EW-01728-65 Mfr # 3922
COMPARE
Flat
$627.00USD / Case of 100
Corning 3915 96-Well Black Solid Plates, Nontreated EW-01728-66 Mfr # 3915
COMPARE
Untreated Flat
$586.00USD / Case of 100
Corning 3925 96-Well Black Solid Plates, High binding EW-01728-67 Mfr # 3925
COMPARE
Flat
$725.00USD / Case of 100