Corning® Conical-Bottom Disposable Plastic Tubes
Ideal for applications where other tubes would shatter
  • Sterile
  • Certified nonpyrogenic
Corning® Tubes are available in either clear polypropylene (PP) or polyethylene terephthalate (PET). Choose either plug seal or flat top caps. The 15- and 50-mL tubes feature black printed graduations and white writing patch. The 50-mL tube 17400-44 is self-standing. The 250- and 500-mL tubes feature molded-in graduations and require an adapter (sold separately below).
$254.75 - $536.00USD / Case of 500
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
Corning 430055 Conical-bottom Disposable Plastic Tubes; 15 mL; Ms; 10 Racks Of 50
Buy More Save More
EW-17300-20 Mfr # 430055
COMPARE
Conical 15 Polyethylene terephthalate (PET)
$387.75USD / Case of 500
Corning 430053 Conical-Bottom Disposable Plastic Tubes; 15 mL; MS; 20 bags of 25
Buy More Save More
EW-17300-22 Mfr # 430053
COMPARE
Conical 15 Modified polystyrene
$377.25USD / Case of 500
Corning 430791 Conical-Bottom Tubes, 15 mL/PP 25x20 Bags
Buy More Save More
EW-17400-22 Mfr # 430791
COMPARE
Conical 15 Polypropylene
$372.75USD / Case of 500
Corning 430052 Conical-Bottom Tubes, 15 mL/PP/Plug; 50x10 Racks
Buy More Save More
EW-17400-24 Mfr # 430052
COMPARE
Conical 15 Polypropylene
$393.00USD / Case of 500
Corning 430766 Conical-Bottom Tubes, 15 mL/PP/Plug; 25x20 Bags
Buy More Save More
EW-17400-26 Mfr # 430766
COMPARE
Conical 15 Polypropylene
$381.50USD / Case of 500
Corning 430828 Conical-Bottom Tubes, 50 mL/PP/Flat; 25x20 Racks
Buy More Save More
EW-17400-30 Mfr # 430828
COMPARE
Conical 50 Polypropylene
$514.50USD / Case of 500
Corning 430290 Conical-Bottom Tubes, 50 mL/PP/Plug; 25x20 Racks
Buy More Save More
EW-17400-34 Mfr # 430290
COMPARE
Conical 50 Polypropylene
$517.00USD / Case of 500
Corning 430921 Conical-Bottom Tubes, 50 mL/PP/Flat; 25x20 Bags
Buy More Save More
EW-17400-44 Mfr # 430921
COMPARE
Conical 50 Polypropylene
$536.00USD / Case of 500
Corning 430776 Conical-Bottom Disposable Plastic Tubes, 250 mL, PP
Buy More Save More
EW-17410-20 Mfr # 430776
COMPARE
Conical 250 Polypropylene
$343.00USD / Case of 102
Corning 431123 Conical-Bottom Disposable Plastic Tubes, 500 mL, PP
Buy More Save More
EW-17410-30 Mfr # 431123
COMPARE
Conical 500 Polypropylene
$254.75USD / Case of 36
Showing 1- 10 of 10