Corning® Sterile Tissue Culture Roller Bottles, Polystyrene
Seamless, one-piece construction reduces risk of breakage
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
Optically-clear polystyrene roller bottles are sterilized by gamma radiation and certified nonpyrogenic. Bottles are tissue culture treated for optimal cell attachment. Bottles are constructed from durable polystyrene and caps are made of HDPE. Models 01836-05 and -06 feature a ribbed design that doubles the growth area without increasing bottle size. Two window areas provide microscopic viewing as well as pouring surfaces.
Choose from three cap type options. The plug seal provides a liquid-tight seal and the Easy grip cap is designed for easy opening. The Easy grip vent cap adds a 0.2 μm nonwetting (hydrophobic) membrane sealed to the cap.
$440.00 - $1,030.00USD / Case of 40
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Closure Culture Area (cm²)
Availability Pricing
Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Plug Seal, 490 cm2; 40/Cs EW-01836-00 Mfr # 430195
COMPARE
150 Plug Seal 490
$440.00USD / Case of 40
Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Easy Grip Cap, 850 cm2; 40/Cs EW-01836-01 Mfr # 430849
COMPARE
255 Easy Grip Cap 850
$605.00USD / Case of 40
Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Easy Grip Vent Cap, 850 cm2; 40/Cs EW-01836-02 Mfr # 431198
COMPARE
255 Easy Grip Vent Cap 850
$730.00USD / Case of 40
Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Easy Grip Cap, 1700 cm2; 40/Cs EW-01836-05 Mfr # 430852
COMPARE
510 Easy Grip Cap 1700
$1,030.00USD / Case of 40
Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Easy Grip Vent Cap, 1700 cm2; 20/Cs EW-01836-06 Mfr # 431191
COMPARE
510 Easy Grip Vent Cap 1700
$595.00USD / Case of 20