Corning Round Non-Treated Cell Culture Dishes
Vented lid design provides consistent gas exchange
  • Optically clear for easily viewing cells
These polystyrene cell culture dishes feature stacking beads for easier handling. Ideal for applications where cell attachment is not required. Sterilized by gamma radiation and certified nonpyrogenic.
$279.00 - $438.00USD / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Corning 430589 cell culture dish, non-treated, 60 mm diameter, 500/cs EW-01835-08 Mfr # 430589
COMPARE
6.3
$279.00USD / Case of 500
Corning 430591 cell culture dish, non-treated, 100 mm diameter, 500/cs EW-01835-09 Mfr # 430591
COMPARE
15
$438.00USD / Case of 500