Corning® Sterile Culture Tubes, Polystyrene
Sterilized tubes save time and reduce cross contamination
  • Manufactured from optically clear polystyrene
  • Threaded screw caps prevent leakage
  • Sterilized by gamma radiation and nonpyrogenic
These tubes are manufactured from optically clear polystyrene and supplied with threaded plug seal caps to prevent leakage. Treated tubes have a hydrophilic and ionic surface that allows optimal cell attachment. Untreated tubes have hydrophobic surface that and can significantly reduce the attachment of most cells. Sterilized by gamma radiation and certified nonpyrogenic.
$730.00 - $750.00USD / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Packaging Tube Capacity (mL) SurfaceTreatment
Availability Pricing
Corning Culture Tubes, Sterile, Polystyrene, Non-Treated, 15 mL, 16 x 125 mm; 500/Cs EW-34534-30 Mfr # 430157
COMPARE
25 Tubes/Pack, 20 Packs/Case 15 Not Treated
$730.00USD / Case of 500
Corning Culture Tubes, Sterile, Polystyrene, Treated, 15 mL, 16 x 125 mm; 500/Cs EW-34534-32 Mfr # 430172
COMPARE
50 Tubes/Pack, 10 Packs/Case 15 Tissue Culture Treated
$750.00USD / Case of 500