Costar Multi-Well Cell Culture Plates with 96-wells
Optically clear, virgin polystyrene is treated for optimal cell attachment
  • Stacking beads and uniform footprints (86 x 128 mm) facilitate stacking and handling
  • Serrations on plate flange/skirt enable you to easily distinguish plates from lids when handling stacks of clusters
  • Lids feature individual condensation rings to reduce well-to-well contamination
  • Gamma radiation sterilized and certified pyrogen-free
$348.00 - $586.00USD / Case of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Sterile Surface Treatment Well Shape
Availability Pricing
Costar 3585 96-well cell culture plates with lid, flat well, treated, sterile, 50/cs EW-01959-20 Mfr # 3585
COMPARE
Yes Cell Culture Flat
$370.00USD / Case of 50
Costar 3596 96-well cell culture plates with lid, flat well, treated, sterile, 50/cs EW-01959-22 Mfr # 3596
COMPARE
Yes Cell Culture Flat
$385.00USD / Case of 50
Costar 3997 96-well cell culture plates with lid, flat well, treated, sterile, 50/cs EW-01959-24 Mfr # 3997
COMPARE
Yes Cell Culture Flat
$348.00USD / Case of 50
Costar 3598 96-well cell culture plates with lid, flat well, treated, sterile, 100/cs EW-01959-26 Mfr # 3598
COMPARE
Yes Cell Culture Flat
$586.00USD / Case of 100
Costar 3799 96-well cell culture plates with lid, round well, treated, sterile, 50/cs EW-01959-28 Mfr # 3799
COMPARE
Yes Cell Culture Round
$412.00USD / Case of 50