Coupling Hardware 4894
Create your own motor mounting with the hardware below
Order one coupling with the same diameter as the motor driving shaft, one coupling with the same diameter as the driven shaft, and one spider (see accessories).
$28.00 - $31.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Motor hp
Availability Pricing
COMPARE
7.2
$28.00USD / Each
COMPARE
7.2
$28.00USD / Each
COMPARE
7.2
$31.00USD / Each
COMPARE
7.2
$28.00USD / Each
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat