This product is now discontinued, View Related Products
Couplings and tube end fittings
Use these fittings with tubing ranging from 1 to 11 mm OD. All standard fittings except models 06473-07 and -08 come with polypropylene (PP) caps, two large VITON O-rings per cap to connect 2 to 4 mm OD tubing, one small VITON O-ring per cap (use together with large O-rings) to connect 1 to 2 mm OD tubing, and one PTFE cone per cap to make chemically inert connections for 1/16" OD tubing or to create zero dead volume fittings for connection to high-pressure fittings (sold below).
Standard fittings 06473-07 and -08 include one fluoroelastomer O-ring per PTFE cap. All chemically inert fittings come with TEFZEL® caps and 1/16" tube OD PTFE cones for improved chemical resistance. All O-rings are made of fluoroelastomer. Max fluid pressure (up to 50 psi) will vary depending on tubing material and size.
Call For Price
35 variations of this product are available.
See All Product Options Below
35 PRODUCT OPTIONS
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Diba Tube end fittings for 1/16", black polypropylene, pack of 10 EW-06471-20 Mfr # 002100
COMPARE
1/16 Polypropylene Tube end fittings for 1/16\", black polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/16", brown polypropylene, pack of 10 EW-06471-21 Mfr # 002101
COMPARE
1/16 Polypropylene Tube End Fittings, for 1/16\", brown polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/16", red polypropylene, pack of 10 EW-06471-22 Mfr # 002102
COMPARE
1/16 Polypropylene Tube end fittings for 1/16\", red polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/16", orange polypropylene, pack of 10 EW-06471-23 Mfr # 002103
COMPARE
1/16 Polypropylene Tube End Fittings, for 1/16\", orange polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/16", yellow polypropylene, pack of 10 EW-06471-24 Mfr # 002104
COMPARE
Polypropylene Tube End Fittings, for 1/16\", yellow polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/16", green polypropylene, pack of 10 EW-06471-25 Mfr # 002105
COMPARE
1/16 Polypropylene Tube End Fittings, for 1/16\", green polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/16", purple polypropylene, pack of 10 EW-06471-27 Mfr # 002107
COMPARE
1/16 Polypropylene Tube End Fittings, for 1/16\", purple polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/16", gray polypropylene, pack of 10 EW-06471-28 Mfr # 002108
COMPARE
1/16 Polypropylene Tube end fittings for 1/16\", gray polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/16", white polypropylene, pack of 10 EW-06471-29 Mfr # 002109
COMPARE
1/16 Polypropylene Tube end fittings for 1/16\", white polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/16", mixed colors, polypropylene, pack of 10 EW-06471-30 Mfr # 002110
COMPARE
1/16 Polypropylene Tube end fittings for 1/16\", mixed colors, polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/8", black polypropylene, pack of 10 EW-06471-40 Mfr # 002200
COMPARE
1/8 Polypropylene Tube end fittings for 1/8\", black polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/8", brown polypropylene, pack of 10 EW-06471-41 Mfr # 002201
COMPARE
1/8 Polypropylene Tube end fittings for 1/8\", brown polypropylene, pack of 10
Call For Price
Diba Tube-end fittings, 1/8", red polypropylene, pack of 10 EW-06471-42 Mfr # 002202
COMPARE
1/8 Polypropylene Tube-end fittings, 1/8\", red polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/8", orange polypropylene, pack of 10 EW-06471-43 Mfr # 002203
COMPARE
Polypropylene Tube end fittings for 1/8\", orange polypropylene, pack of 10
Call For Price
Diba Tube end fittings for 1/8", yellow polypropylene, pack of 10 EW-06471-44 Mfr # 002204
COMPARE
1/8 Polypropylene Tube end fittings for 1/8\", yellow polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/8", green polypropylene, pack of 10 EW-06471-45 Mfr # 002205
COMPARE
1/8 Polypropylene Tube End Fittings, for 1/8\", green polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/8", blue polypropylene, pack of 10 EW-06471-46 Mfr # 002206
COMPARE
1/8 Polypropylene Tube End Fittings, for 1/8\", blue polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/8", purple polypropylene, pack of 10 EW-06471-47 Mfr # 002207
COMPARE
1/8 Polypropylene Tube End Fittings, for 1/8\", purple polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/8", gray polypropylene, pack of 10 EW-06471-48 Mfr # 002208
COMPARE
1/8 Polypropylene Tube End Fittings, for 1/8\", gray polypropylene, pack of 10
Call For Price
Diba Tube End Fittings, for 1/8", white polypropylene, pack of 10 EW-06471-49 Mfr # 002209
COMPARE
1/8 Polypropylene Tube End Fittings, for 1/8\", white polypropylene, pack of 10
Call For Price
Showing 1 - 20 of 35