CPC® AseptiQuik® G Series Genderless Quick-Disconnect Fittings
Provide easy, secure sterile connections even in nonsterile environments
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. The easy three-step FLIP-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. The audible “click” confirms a securely connected assembly.
All fittings can be sterilized by gamma irradiation. Use high-temperature fittings to sterilize via autoclave.
$205.00 - $255.00USD / Pkg of 5
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Sterilization Method Description
Availability Pricing
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, 1/4" ID; 5/Pk
New Products
EW-20202-40 Mfr # AQG17004
COMPARE
1/4 Gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, 1/4" ID; 5/Pk
$205.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, High Temperature, 1/4" ID; 5/Pk
New Products
EW-20202-41 Mfr # AQG17004HT
COMPARE
1/4 Autoclave or gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, High Temperature, 1/4" ID; 5/Pk
$250.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, 3/8" ID; 5/Pk
New Products
EW-20202-42 Mfr # AQG17006
COMPARE
3/8 Gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, 3/8" ID; 5/Pk
$205.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, High Temperature, 3/8" ID; 5/Pk
New Products
EW-20202-43 Mfr # AQG17006HT
COMPARE
3/8 Autoclave or gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, High Temperature, 3/8" ID; 5/Pk
$250.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, 1/2" ID; 5/Pk
New Products
EW-20202-44 Mfr # AQG17008
COMPARE
1/2 Gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, 1/2" ID; 5/Pk
$205.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, High Temperature, 1/2" ID; 5/Pk
New Products
EW-20202-45 Mfr # AQG17008HT
COMPARE
1/2 Autoclave or gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, High Temperature, 1/2" ID; 5/Pk
$250.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, 3/4" ID; 5/Pk
New Products
EW-20202-46 Mfr # AQG17012
COMPARE
3/4 Gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, 3/4" ID; 5/Pk
$215.00USD / Pkg of 5
CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, High Temperature, 3/4" ID; 5/Pk
New Products
EW-20202-47 Mfr # AQG17012HT
COMPARE
3/4 Autoclave or gamma AseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, High Temperature, 3/4" ID; 5/Pk
$255.00USD / Pkg of 5
Showing 1- 8 of 8