CPC® AseptiQuik-S® Hose Barb Connectors, Low Flow
Provide easy, secure sterile connections even in nonsterile environments
  • Quick, three-step connection process reduces risk of operator error
  • Large housing size allows easy installation
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. Push the connectors together; an audible snap confirms a tight connection. Align and snap membrane connectors together and twist blue lock ring clockwise to hear another audible click. Alignment of the ring rib with the body’s arrow indicator confirms final, secure connection.
All fittings can be sterilized by gamma irradiation. Use high temperature fittings to sterilize via autoclave.
$47.00 - $47.75USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Description
Availability Pricing
CPC (Colder) AseptiQuik-S® Hose Barb Connector, Low Flow, High Temperature; 1/8" ID EW-20002-92 Mfr # AQS17002HT
COMPARE
Quick-Disconnect x Barb AseptiQuik-S® Hose Barb Connector, Low Flow, High Temperature; 1/8" ID
$47.00USD / Each
CPC (Colder) AseptiQuik-S® Hose Barb Connector, Low Flow, High Temperature; 1/4" ID EW-20002-93 Mfr # AQS17004HT
COMPARE
Quick-Disconnect x Barb AseptiQuik-S® Hose Barb Connector, Low Flow, High Temperature; 1/4" ID
$47.00USD / Each
CPC (Colder) AseptiQuik-S® Hosebarb Connector, Low Flow High Temperature; 3/4" Mini Tri-Clamp EW-20002-96 Mfr # AQS33012HT
COMPARE
Quick-Disconnect x Sanitary AseptiQuik-S® Hose Barb Connector, Low Flow, High Temperature; 3/4" Mini Tri-Clamp
$47.75USD / Each