CPC® High-Flow Quick-Disconnect Fittings, Bulkhead Panel Mount Hose Barb Bodies
No more leaks—valved couplings automatically shut off flow
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.
$19.70 - $325.00USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Valved, 3/8" ID
Buy More Save More
EW-31303-20 Mfr # HFCD16612
COMPARE
3/8 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Valved, 3/8" ID; 1/Ea
$24.25USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Valved, 1/2" ID
Buy More Save More
EW-31303-21 Mfr # HFCD16812
COMPARE
1/2 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Valved, 1/2" ID; 1/Ea
$24.25USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 1/2" ID
Buy More Save More
EW-31303-26 Mfr # HFC16812
COMPARE
1/2 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 1/2" ID; 1/Ea
$19.70USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 3/4" ID
Buy More Save More
EW-31303-28 Mfr # HFC161212
COMPARE
3/4 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 3/4" ID; 1/Ea
$22.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Valved, 3/8" ID
Buy More Save More
EW-31307-20 Mfr # HFCD16635
COMPARE
3/8 Polysulfone High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Valved, 3/8" ID; 1/Ea
$31.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Valved, 3/4" ID; 10/Pk EW-31308-36 Mfr # HFCD161212
COMPARE
3/4 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Valved, 3/4" ID; 10/Pk
$227.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 3/8" ID; 10/Pk EW-31308-37 Mfr # HFC16612
COMPARE
3/8 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 3/8" ID; 10/Pk
$218.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 5/8" ID; 10/Pk EW-31308-38 Mfr # HFC161012
COMPARE
5/8 Polypropylene High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polypropylene, Straight-Through, 5/8" ID; 10/Pk
$218.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Valved, 1/2" ID; 10/Pk EW-31308-65 Mfr # HFCD16835
COMPARE
1/2 Polysulfone High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Valved, 1/2" ID; 10/Pk
$273.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Valved, 3/4" ID; 10/Pk EW-31308-67 Mfr # HFCD161235
COMPARE
3/4 Polysulfone High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Valved, 3/4" ID; 10/Pk
$325.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Straight-Through, 3/8" ID; 10/Pk EW-31308-68 Mfr # HFC16635
COMPARE
3/8 Polysulfone High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Straight-Through, 3/8" ID; 10/Pk
$213.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Straight-Through, 1/2" ID; 10/Pk EW-31308-69 Mfr # HFC16835
COMPARE
1/2 Polysulfone High-Flow Quick-Disconnect Fitting, Bulkhead Panel Mount Hose Barb Body, Polysulfone, Straight-Through, 1/2" ID; 10/Pk
$223.00USD / Pkg of 10
Showing 1- 12 of 12