CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
No more leaks—valved couplings automatically shut off flow
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.
$19.00 - $242.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Valved, 3/8" ID
Buy More Save More
EW-31303-10 Mfr # HFCD17612
COMPARE
3/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 3/8" ID; 1/Ea
$21.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Valved, 1/2" ID
Buy More Save More
EW-31303-11 Mfr # HFCD17812
COMPARE
1/2 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 1/2" ID; 1/Ea
$23.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Valved, 5/8" ID
Buy More Save More
EW-31303-12 Mfr # HFCD171012
COMPARE
5/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 5/8" ID; 1/Ea
$21.50USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Valved, 3/4" ID
Buy More Save More
EW-31303-13 Mfr # HFCD171212
COMPARE
3/4 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 3/4" ID; 1/Ea
$21.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Straight-Through, 3/8" ID
Buy More Save More
EW-31303-15 Mfr # HFC17612
COMPARE
3/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Straight-Through, 3/8" ID; 1/Ea
$19.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Straight-Through, 1/2" ID
Buy More Save More
EW-31303-16 Mfr # HFC17812
COMPARE
1/2 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Straight-Through, 1/2" ID; 1/Ea
$19.20USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Straight-Through,3/4" ID
Buy More Save More
EW-31303-18 Mfr # HFC171212
COMPARE
3/4 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Straight-Through,3/4" ID; 1/Ea
$20.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 3/8" ID
Buy More Save More
EW-31307-10 Mfr # HFCD17635
COMPARE
3/8 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 3/8" ID; 1/Ea
$28.50USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 1/2" ID
Buy More Save More
EW-31307-11 Mfr # HFCD17835
COMPARE
1/2 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 1/2" ID; 1/Ea
$25.50USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 3/4" ID
Buy More Save More
EW-31307-13 Mfr # HFCD171235
COMPARE
3/4 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 3/4" ID; 1/Ea
$25.75USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polysulfone, Straight-Through, 3/8" ID
Buy More Save More
EW-31307-15 Mfr # HFC17635
COMPARE
3/8 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Straight-Through, 3/8" ID; 1/Ea
$22.50USD / Each
CPC (Colder) HFC171235 Quick-disconnect fittings, Polysulfone straight-through hose barbs, 3/4"
Buy More Save More
EW-31307-18 Mfr # HFC171235
COMPARE
3/4 Polysulfone Quick-disconnect fittings, Polysulfone straight-through hose barbs, 3/4"
$21.80USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Straight-Through, 5/8" ID; 10/Pk EW-31308-34 Mfr # HFC171012
COMPARE
5/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Straight-Through, 5/8" ID; 10/Pk
$182.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polysulfone, Valved, 5/8" ID; 10/Pk EW-31308-62 Mfr # HFCD171035
COMPARE
5/8 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 5/8" ID; 10/Pk
$242.00USD / Pkg of 10
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Body, Polysulfone, Straight-Through, 1/2" ID; 10/Pk EW-31308-63 Mfr # HFC17835
COMPARE
1/2 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Straight-Through, 1/2" ID; 10/Pk
$204.00USD / Pkg of 10
Showing 1- 15 of 15