CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
No more leaks—valved couplings automatically shut off flow
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.
$8.70 - $104.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 3/8" ID
Buy More Save More
EW-31303-40 Mfr # HFC22612
COMPARE
3/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 3/8" ID; 1/Ea
$9.20USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/2" ID
Buy More Save More
EW-31303-41 Mfr # HFC22812
COMPARE
1/2 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/2" ID; 1/Ea
$8.70USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 5/8" ID
Buy More Save More
EW-31303-42 Mfr # HFC221012
COMPARE
5/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 5/8" ID; 1/Ea
$9.60USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 3/4" ID
Buy More Save More
EW-31303-43 Mfr # HFC221212
COMPARE
3/4 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 3/4" ID; 1/Ea
$9.90USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polypropylene, Valved, 3/8" ID
Buy More Save More
EW-31303-45 Mfr # HFCD22612
COMPARE
3/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 3/8" ID; 1/Ea
$18.40USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polypropylene, Valved, 1/2" ID
Buy More Save More
EW-31303-46 Mfr # HFCD22812
COMPARE
1/2 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 1/2" ID; 1/Ea
$16.70USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polypropylene, Valved, 5/8" ID
Buy More Save More
EW-31303-47 Mfr # HFCD221012
COMPARE
5/8 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 5/8" ID; 1/Ea
$17.80USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polypropylene, Valved, 3/4" ID
Buy More Save More
EW-31303-48 Mfr # HFCD221212
COMPARE
3/4 Polypropylene High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 3/4" ID; 1/Ea
$17.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Straight-Through, 3/8" ID
Buy More Save More
EW-31307-40 Mfr # HFC22635
COMPARE
3/8 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Straight-Through, 3/8" ID; 1/Ea
$10.90USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Straight-Through, 1/2" ID
Buy More Save More
EW-31307-41 Mfr # HFC22835
COMPARE
1/2 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Straight-Through, 1/2" ID; 1/Ea
$11.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Valved, 3/8" ID
Buy More Save More
EW-31307-45 Mfr # HFCD22635
COMPARE
3/8 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, 3/8" ID; 1/Ea
$22.25USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Valved, 1/2" ID
Buy More Save More
EW-31307-46 Mfr # HFCD22835
COMPARE
1/2 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, 1/2" ID; 1/Ea
$21.00USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Valved, 5/8" ID
Buy More Save More
EW-31307-47 Mfr # HFCD221035
COMPARE
5/8 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, 5/8" ID; 1/Ea
$20.50USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Valved, 3/4" ID
Buy More Save More
EW-31307-48 Mfr # HFCD221235
COMPARE
3/4 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, 3/4" ID; 1/Ea
$21.75USD / Each
CPC (Colder) High-Flow Quick-Disconnect Fitting, Hosebarb Insert, Polysulfone, Straight-Through, 3/4" ID; 10/Pk EW-31308-74 Mfr # HFC221235
COMPARE
3/4 Polysulfone High-Flow Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Straight-Through, 3/4" ID; 10/Pk
$104.00USD / Pkg of 10
Showing 1- 15 of 15