CPC® Metal Quick-Disconnect Fittings, NPT (M) Threaded Inserts
Convenience of a quick-disconnect in pressure and vacuum applications
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).
$9.90 - $25.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Quick-Disconnect Flow Size (in) Thread size NPT(M) (port 1) (in) Description
Availability Pricing
CPC (Colder) LCD24004 Metal Quick-Disconnect Fitting, NPT (M) Threaded Insert, Valved, 1/4" NPT(M)
Buy More Save More
EW-31403-50 Mfr # LCD24004
COMPARE
1/4 1/4 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Valved, 1/4" NPT(M); 1/Ea
$23.75USD / Each
CPC (Colder) Metal Quick-Disconnect Fitting, NPT (M) Threaded Insert, Valved, 3/8" NPT(M)
Buy More Save More
EW-31403-51 Mfr # LCD24006
COMPARE
1/4 3/8 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Valved, 3/8" NPT(M); 1/Ea
$25.00USD / Each
CPC (Colder) LC Series Metal Quick-Disconnect Fitting, NPT (M) Thread Insert, Straight-Through, 1/4" NPT(M)
Buy More Save More
EW-31403-55 Mfr # LC24004
COMPARE
1/4 1/4 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Straight-Through, 1/4" NPT(M); 1/Ea
$10.20USD / Each
CPC (Colder) Metal Quick-Disconnect Fitting, NPT (M) Thread Insert, Straight-Through, 3/8" NPT(M)
Buy More Save More
EW-31403-56 Mfr # LC24006
COMPARE
1/4 3/8 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Straight-Through, 3/8" NPT(M); 1/Ea
$15.50USD / Each
CPC (Colder) Metal Quick-Disconnect Fitting, NPT (M) Thread Insert, Straight-Through, 1/8" NPT(M)
Buy More Save More
EW-31404-40 Mfr # MC2402
COMPARE
1/8 1/8 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Straight-Through, 1/8" NPT(M); 1/Ea
$9.90USD / Each
CPC (Colder) MC Series Metal Quick-Disconnect Fitting, NPT (M) Thread Insert, Straight-Through, 1/4" NPT(M)
Buy More Save More
EW-31404-41 Mfr # MC2404
COMPARE
1/8 1/4 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Straight-Through, 1/4" NPT(M); 1/Ea
$10.70USD / Each
CPC (Colder) Metal Quick-Disconnect Fitting, NPT (M) Threaded Insert, Valved, 1/8" NPT(M)
Buy More Save More
EW-31404-60 Mfr # MCD2402
COMPARE
1/8 1/8 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Valved, 1/8" NPT(M); 1/Ea
$20.50USD / Each
CPC (Colder) MCD2404 Metal Quick-Disconnect Fitting, NPT (M) Threaded Insert, Valved, 1/4" NPT(M)
Buy More Save More
EW-31404-61 Mfr # MCD2404
COMPARE
1/8 1/4 Metal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Valved, 1/4" NPT(M); 1/Ea
$19.10USD / Each
Showing 1- 8 of 8