CPC® Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body
Convenience of a quick-disconnect in pressure and vacuum applications
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).
$27.75 - $247.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Quick-Disconnect Flow Size (in) Description
Availability Pricing
CPC (Colder) Metal Quick-Disconnect Fitting, Panel-Mount Thread Body, Valved, 1/8" NPT(M); 10/Pk EW-31309-33 Mfr # MCD1502
COMPARE
Quick-Disconnect x NPT(M) 1/8 Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Valved, 1/8" NPT(M); 10/Pk
$234.00USD / Pkg of 10
CPC (Colder) Metal Quick-Disconnect Fitting, Panel-Mount Thread Body, Straight-Through, 1/8" NPT(M); 10/Pk EW-31309-34 Mfr # MC1502
COMPARE
Quick-Disconnect x NPT(M) 1/8 Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Straight-Through, 1/8" NPT(M); 10/Pk
$242.00USD / Pkg of 10
CPC (Colder) Metal Quick-Disconnect Fitting, Panel-Mount Thread Body, Straight-Through, 1/4" NPT(M); 10/Pk EW-31309-35 Mfr # LC15004
COMPARE
Quick-Disconnect x NPT(M) 1/4 Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Straight-Through, 1/4" NPT(M); 10/Pk
$221.00USD / Pkg of 10
CPC (Colder) Metal Quick-Disconnect Fitting, Panel-Mount Thread Body, Straight-Through, 10-32 UNF; 10/Pk EW-31309-36 Mfr # MC181032
COMPARE
Quick-Disconnect x UNF(M) 1/8 Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Straight-Through, 10-32 UNF; 10/Pk
$247.00USD / Pkg of 10
CPC (Colder) Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Valved, 1/4" NPT(M)
Buy More Save More
EW-31413-14 Mfr # LCD15004
COMPARE
Quick-Disconnect x NPT(M) 1/4 Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Valved, 1/4" NPT(M); 1/Ea
$27.75USD / Each
CPC (Colder) Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Valved, 10-32 UNF
Buy More Save More
EW-31413-15 Mfr # MCD181032
COMPARE
Quick-Disconnect x UNF(M) 1/8 Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body, Valved, 10-32 UNF; 1/Ea
$31.50USD / Each