CPC® Metal Quick-Disconnect Fittings, Threaded Bodies
Convenience of a quick-disconnect in pressure and vacuum applications
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).
$17.40 - $24.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Quick-Disconnect Flow Size (in) Thread size NPT(M) (port 1) (in) Description
Availability Pricing
CPC (Colder) LCD10004 Metal Quick-Disconnect Fittings, Threaded Body, Valved, 1/4" NPT(M)
Buy More Save More
EW-31403-00 Mfr # LCD10004
COMPARE
1/4 1/4 Metal Quick-Disconnect Fittings, Threaded Body, Valved, 1/4" NPT(M); 1/Ea
$24.50USD / Each
CPC (Colder) Metal Quick-Disconnect Fittings, Threaded Body, Valved, 3/8" NPT(M)
Buy More Save More
EW-31403-01 Mfr # LCD10006
COMPARE
1/4 3/8 Metal Quick-Disconnect Fittings, Threaded Body, Valved, 3/8" NPT(M); 1/Ea
$23.25USD / Each
CPC (Colder) LC10004 Metal Quick-Disconnect Fitting, Threaded Body, Straight-Through, 1/4" NPT(M)
Buy More Save More
EW-31403-05 Mfr # LC10004
COMPARE
1/4 1/4 Metal Quick-Disconnect Fittings, Threaded Body, Straight-Through, 1/4" NPT(M); 1/Ea
$22.25USD / Each
CPC (Colder) LC10006 Metal Quick-Disconnect Fitting, Threaded Body, Straight-Through, 1/4" NPT(M)
Buy More Save More
EW-31403-06 Mfr # LC10006
COMPARE
1/4 3/8 Metal Quick-Disconnect Fittings, Threaded Body, Straight-Through, 1/4" NPT(M); 1/Ea
$23.00USD / Each
CPC (Colder) Metal Quick-Disconnect Fitting, Threaded Body, Straight-Through, 1/8" NPT(M)
Buy More Save More
EW-31404-00 Mfr # MC1002
COMPARE
1/8 1/8 Metal Quick-Disconnect Fittings, Threaded Body, Straight-Through, 1/8" NPT(M); 1/Ea
$17.90USD / Each
CPC (Colder) MC1004 Metal Quick-Disconnect Fitting, Threaded Body, Straight-Through, 1/4" NPT(M)
Buy More Save More
EW-31404-01 Mfr # MC1004
COMPARE
1/8 1/4 Metal Quick-Disconnect Fittings, Threaded Body, Straight-Through, 1/4" NPT(M); 1/Ea
$17.40USD / Each
CPC (Colder) Metal Quick-Disconnect Fittings, Threaded Body, Valved, 1/8" NPT(M)
Buy More Save More
EW-31404-20 Mfr # MCD1002
COMPARE
1/8 1/8 Metal Quick-Disconnect Fittings, Threaded Body, Valved, 1/8" NPT(M); 1/Ea
$19.10USD / Each
CPC (Colder) MCD1004 Metal Quick-Disconnect Fitting, Threaded Body, Valved, 1/4" NPT(M)
Buy More Save More
EW-31404-21 Mfr # MCD1004
COMPARE
1/8 1/4 Metal Quick-Disconnect Fittings, Threaded Body, Valved, 1/4" NPT(M); 1/Ea
$19.10USD / Each
Showing 1- 8 of 8