CPC® Miniature Quick-Disconnect Fittings, Hose Barb Inserts
No more kinked tubing—allows tubing to rotate freely
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected—preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.
$23.00 - $943.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) Miniature Quick-Disconnect Fitting, Hosebarb Insert, Acetal, Straight-Through, 1/16" ID; 10/Pk
Buy More Save More
EW-06363-50 Mfr # SMM01
COMPARE
1/16 Acetal Miniature Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Straight-Through, 1/16" ID; 10/Pk
$39.75USD / Pkg of 10
CPC (Colder) Miniature Quick-Disconnect Fitting, Hosebarb Insert, Acetal,Straight-Through, 1/8" ID; 10/Pk
Buy More Save More
EW-06363-52 Mfr # SMM02
COMPARE
1/8 Acetal Miniature Quick-Disconnect Fitting, Hose Barb Insert, Acetal,Straight-Through, 1/8" ID; 10/Pk
$23.00USD / Pkg of 10
CPC (Colder) Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/16" ID; 10/Pk
Buy More Save More
EW-06363-58 Mfr # SMM0112
COMPARE
1/16 Polypropylene Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/16" ID; 10/Pk
$34.50USD / Pkg of 10
CPC (Colder) Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/8" ID; 10/Pk
Buy More Save More
EW-06363-60 Mfr # SMM0212
COMPARE
1/8 Polypropylene Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/8" ID; 10/Pk
$33.50USD / Pkg of 10
CPC (Colder) Miniature Quick-Disconnect Fitting, Hosebarb Insert, Acetal,Straight-Through, 3 mm ID; 10/Pk
Buy More Save More
EW-06363-65 Mfr # SMMM3
COMPARE
1/8 Acetal Miniature Quick-Disconnect Fitting, Hose Barb Insert, Acetal,Straight-Through, 3 mm ID; 10/Pk
$741.00USD / Pkg of 10
CPC (Colder) Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Straight-Through, 1/8" ID; 10/Pk
Buy More Save More
EW-06363-84 Mfr # SMM0291
COMPARE
1/8 Polycarbonate Miniature Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Straight-Through, 1/8" ID; 10/Pk
$943.00USD / Pkg of 10