CPC® Plastic Quick-Disconnect Fittings, Complete Couplings
Fittings connect in a snap
  • Acetal fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
Complete couplings include one fitting insert and one fitting body.
$79.00 - $204.00USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Complete Hose Barb Coupling, Acetal, 1/4" ID; 10/Pk
Buy More Save More
EW-06360-90 Mfr # APC04
COMPARE
1/4 1/4 Acetal
$171.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting, Complete Hose Barb Coupling, Acetal, 3/8" ID; 10/Pk
Buy More Save More
EW-06360-91 Mfr # APC06
COMPARE
3/8 3/8 Acetal
$204.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting, Complete Hosebarb Coupling, Polycarbonate, 1/4" ID; 10/Pk
Buy More Save More
EW-31305-00 Mfr # 1044200
COMPARE
1/4 1/4 Polycarbonate
$79.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting, Complete Hosebarb Coupling, Polycarbonate, 3/8" ID; 10/Pk
Buy More Save More
EW-31305-05 Mfr # 1044300
COMPARE
3/8 3/8 Polycarbonate
$79.50USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting, Complete Hosebarb Coupling, Polysulfone, 3/8" ID; 10/Pk
Buy More Save More
EW-31306-05 Mfr # 1044100
COMPARE
3/8 3/8 Polysulfone
$123.00USD / Pkg of 10