CPC® Plastic Quick-Disconnect Fittings, Sealing Caps
Fittings connect in a snap
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).
$20.50 - $4,443.00USD / Pkg of 5
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Quick-Disconnect Flow Size (in) Description
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Sealing Cap, Polycarnonate, 1/4" Flow Size; 5/Pk
Buy More Save More
EW-31311-64 Mfr # MPC32003
COMPARE
Polycarbonate 1/4 Plastic Quick-Disconnect Fitting, Sealing Cap, Polycarbonate, 1/4" Flow Size; 5/Pk
$20.50USD / Pkg of 5
CPC (Colder) Quick-Disconnect Fitting, Sealing Cap, Polycarnonate, 1/2" Flow Size; 5/Pk
Buy More Save More
EW-31311-65 Mfr # MPX32003
COMPARE
Polycarbonate 1/2 Plastic Quick-Disconnect Fitting, Sealing Cap, Polycarbonate, 1/2" Flow Size; 5/Pk
$34.25USD / Pkg of 5
CPC (Colder) Quick-Disconnect Fitting, Sealing Cap with Lock, Polycarnonate, 1/4" Flow Size; 5/Pk
Buy More Save More
EW-31311-67 Mfr # MPCK32003
COMPARE
Polycarbonate 1/4 Plastic Quick-Disconnect Fitting, Sealing Cap with Lock, Polycarbonate, 1/4" Flow Size; 5/Pk
$36.00USD / Pkg of 5
CPC (Colder) Plastic Quick-Disconnect Fitting, Sealing Cap, Polysulfone, 1/4" Flow Size; 5/Pk
Buy More Save More
EW-31311-74 Mfr # MPC32039
COMPARE
Polysulfone 1/4 Plastic Quick-Disconnect Fitting, Sealing Cap, Polysulfone, 1/4" Flow Size; 5/Pk
$42.50USD / Pkg of 5
CPC (Colder) Quick-Disconnect Fitting, Sealing Cap with Lock, Polysulfone, 1/4" Flow Size; 5/Pk
Buy More Save More
EW-31311-77 Mfr # MPCK32039
COMPARE
Polysulfone 1/4 Plastic Quick-Disconnect Fitting, Sealing Cap with Lock, Polysulfone, 1/4" Flow Size; 5/Pk
$44.00USD / Pkg of 5
CPC (Colder) Quick-Disconnect Fitting, Sealing Cap, Polysulfone, 3/8" Flow Size; 100/Bg EW-41001-07 Mfr # MPX32039
COMPARE
Polysulfone 3/8 Plastic Quick-Disconnect Fitting, Sealing Cap, Polysulfone, 3/8" Flow Size; 25/Pk
$4,443.00USD / Bag of 100