CPC® Plastic Quick-Disconnect Fittings, Sealing Plugs
Fittings connect in a snap
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).
$18.40 - $36.00USD / Pkg of 5
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Quick-Disconnect Flow Size (in) Description
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 1/4" Flow Size; 5/Pk
Buy More Save More
EW-31311-68 Mfr # MPC30003M
COMPARE
Polycarbonate 1/4 Plastic Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 1/4" Flow Size; 5/Pk
$18.40USD / Pkg of 5
CPC (Colder) Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 3/8" Flow Size; 5/Pk
Buy More Save More
EW-31311-69 Mfr # MPX30003M
COMPARE
Polycarbonate 1/2 Plastic Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 3/8" Flow Size; 5/Pk
$36.00USD / Pkg of 5
CPC (Colder) Quick-Disconnect Fitting, Sealing Plug, Polysulfone, 1/4" Flow Size; 5/Pk
Buy More Save More
EW-31311-78 Mfr # MPC30039M
COMPARE
Polysulfone 1/4 Plastic Quick-Disconnect Fitting, Sealing Plug, Polysulfone, 1/4" Flow Size; 5/Pk
$29.25USD / Pkg of 5