CPC® Quick-Disconnect Fittings, 1⁄4-28 Flat-Bottom Port Bodies
No more struggling—just press thumb latch
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
The 1/4-28 flat-bottom tubing ports with compression nuts provide a secure connection to eliminate worries of tubing line leak at the fitting.
A complete coupler requires a fitting body and an insert (order separately).
$15.40 - $17.20USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Description
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, 1⁄4-28 Flat-Bottom Port Body, Polypropylene, Valved EW-06368-69 Mfr # PMCD19042812
COMPARE
Polypropylene Quick-Disconnect x UNF(F) Quick-Disconnect Fitting, 1⁄4-28 Flat-Bottom Port Body, Polypropylene, Valved; 1/Ea
$15.40USD / Each
CPC (Colder) Quick-Disconnect Fitting, 1⁄4-28 Flat-Bottom Port Body, Panel-Mount, Polypropylene, Valved
Always in Stock
EW-06368-70 Mfr # PMCD18042812
COMPARE
Polypropylene Quick-Disconnect x UNF(F) Quick-Disconnect Fitting, 1⁄4-28 Flat-Bottom Port Body, Panel-Mount, Polypropylene, Valved; 1/Ea
$17.20USD / Each