CPC® Quick-Disconnect Fittings, Compression Bodies
No more struggling—just press thumb latch
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
$10.70 - $479.00USD / Pkg of 25
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Quick-Disconnect Flow Size (in)
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Acetal, Valved, 1/4" Flow Size; 1/4" OD
Buy More Save More
EW-06359-90 Mfr # PLCD13004
COMPARE
1/4 Acetal 1/4
$18.60USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Acetal, Valved, 3/8" OD
Buy More Save More
EW-06359-91 Mfr # PLCD13006
COMPARE
3/8 Acetal 1/4
$17.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Acetal, Valved, 1/8" Flow Size, 1/4" OD
Buy More Save More
EW-06360-05 Mfr # PMCD1204
COMPARE
1/4 Acetal 1/8
$17.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Acetal, Valved, 1/4" Flow Size, 1/4" OD
Buy More Save More
EW-06360-49 Mfr # PLCD12004
COMPARE
1/4 Acetal 1/4
$20.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Acetal, Valved, 3/8" OD
Buy More Save More
EW-06360-50 Mfr # PLCD12006
COMPARE
3/8 Acetal 1/4
$21.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Acetal, Valved, 1/8" Flow Size; 1/4" OD
Buy More Save More
EW-06360-98 Mfr # PMCD1304
COMPARE
1/4 Acetal 1/8
$12.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Acetal, Straight-Through, 1/4" OD
Buy More Save More
EW-06361-02 Mfr # PMC1204
COMPARE
1/4 Acetal 1/8
$16.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Acetal, Straight-Through, 1/4" OD
Buy More Save More
EW-06361-08 Mfr # PMC1304
COMPARE
1/4 Acetal 1/8
$10.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Acetal, Straight-Through, 3/8" OD
Buy More Save More
EW-06361-49 Mfr # PLC13006
COMPARE
3/8 Acetal 1/4
$17.60USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Polypropylene, Valved, 1/4" OD
Buy More Save More
EW-06364-05 Mfr # PMCD120412
COMPARE
1/4 Polypropylene 1/8
$19.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Polypropylene, Valved, 3/8" OD
Buy More Save More
EW-06364-50 Mfr # PLCD1200612
COMPARE
3/8 Polypropylene 1/4
$25.25USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Polypropylene, Valved, 1/4" OD
Buy More Save More
EW-06364-90 Mfr # PMCD130412
COMPARE
1/4 Polypropylene 1/8
$18.40USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Polypropylene, Valved, 3/8" OD
Buy More Save More
EW-06364-91 Mfr # PLCD1300612
COMPARE
3/8 Polypropylene 1/4
$24.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Polypropylene, Valved, 1/4" OD; 25/Pk EW-31307-03 Mfr # PMCD130412
COMPARE
1/4 Polypropylene 1/8
$431.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Acetal, Straight-Through, 1/4" OD; 25/Pk EW-31307-33 Mfr # PLC12004
COMPARE
1/4 Acetal 1/4
$479.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Acetal, Straight-Through, 3/8" OD; 25/Pk EW-31307-34 Mfr # PLC12006
COMPARE
3/8 Acetal 1/4
$475.00USD / Pkg of 25
Showing 1- 16 of 16