CPC® Quick-Disconnect Fittings, Compression Inserts
No more struggling—just press thumb latch
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
$6.70 - $357.00USD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/8" Flow Size, 1/4" OD
Buy More Save More
EW-06360-30 Mfr # PMCD2004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/8" Flow Size, 1/4" OD; 1/Ea
$10.30USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/8" Flow Size, 1/4" OD
Buy More Save More
EW-06360-32 Mfr # PMC2004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/8" Flow Size, 1/4" OD; 1/Ea
$6.90USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/8" Flow Size, 3/8" OD
Buy More Save More
EW-06360-35 Mfr # PMCD2006
COMPARE
3/8 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/8" Flow Size, 3/8" OD; 1/Ea
$14.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/8" Flow Size, 3/8" OD
Buy More Save More
EW-06360-37 Mfr # PMC2006
COMPARE
3/8 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/8" Flow Size, 3/8" OD; 1/Ea
$7.60USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 5/32" OD
Buy More Save More
EW-06360-41 Mfr # PMCD20025
COMPARE
5/32 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 5/32" OD; 1/Ea
$15.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/4" Flow Size, 1/4" OD
Buy More Save More
EW-06360-74 Mfr # PLCD20004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/4" Flow Size, 1/4" OD; 1/Ea
$11.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/4" Flow Size, 3/8" OD
Buy More Save More
EW-06360-75 Mfr # PLCD20006
COMPARE
3/8 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, 1/4" Flow Size, 3/8" OD; 1/Ea
$11.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/4" Flow Size, 1/4" OD
Buy More Save More
EW-06360-76 Mfr # PLC20004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/4" Flow Size, 1/4" OD; 1/Ea
$6.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/4" Flow Size, 3/8" OD
Buy More Save More
EW-06360-77 Mfr # PLC20006
COMPARE
3/8 Acetal Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, 1/4" Flow Size, 3/8" OD; 1/Ea
$7.30USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved; 1/4" OD
Buy More Save More
EW-06361-32 Mfr # PMCD4004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, 1/4" OD; 1/Ea
$16.80USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, 1/4" OD
Buy More Save More
EW-06364-30 Mfr # PMCD200412
COMPARE
1/4 Polypropylene Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, 1/4" OD; 1/Ea
$16.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Polypropylene, Straight-Through, 1/4" OD
Buy More Save More
EW-06364-32 Mfr # PMC200412
COMPARE
1/4 Polypropylene Quick-Disconnect Fitting, Compression Insert, Polypropylene, Straight-Through, 1/4" OD; 1/Ea
$7.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, 3/8" OD
Buy More Save More
EW-06364-75 Mfr # PLCD2000612
COMPARE
3/8 Polypropylene Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, 3/8" OD; 1/Ea
$18.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, 1/8" Flow Size, 1/4" OD; 25/Pk EW-31307-08 Mfr # PMC4004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, 1/8" Flow Size, 1/4" OD; 25/Pk
$263.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, 1/4" Flow Size, 1/4" OD; 25/Pk EW-31307-39 Mfr # PLC40004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, 1/4" Flow Size, 1/4" OD; 25/Pk
$283.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, 3/8" OD; 25/Pk EW-31307-44 Mfr # PLC40006
COMPARE
3/8 Acetal Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, 3/8" OD; 25/Pk
$254.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, 1/4" OD; 25/Pk EW-31307-53 Mfr # PLCD40004
COMPARE
1/4 Acetal Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, 1/4" OD; 25/Pk
$342.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, 3/8" OD; 25/Pk EW-31307-54 Mfr # PLCD40006
COMPARE
3/8 Acetal Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, 3/8" OD; 25/Pk
$357.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Compression Insert; Straight-through, 3/8" Tube OD, PP; 25/PK EW-31307-72 Mfr # PLC2000612
COMPARE
3/8 Polypropylene Quick-Disconnect Compression Insert; Straight-through, 3/8" Tube OD, PP
$177.00USD / Pkg of 25
Showing 1- 19 of 19