CPC® Quick-Disconnect Fittings, Hose Barb Bodies
No more struggling—just press thumb latch
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
$8.60 - $421.00USD / Each
38 variations of this product are available.
See All Product Options Below
38 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Quick-Disconnect Flow Size (in)
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Acetal, Valved, 1/16" ID
Buy More Save More
EW-06360-09 Mfr # PMCD1601
COMPARE
1/16 Acetal 1/8
$17.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Acetal, Valved, 1/8" ID
Buy More Save More
EW-06360-10 Mfr # PMCD1602
COMPARE
1/8 Acetal 1/8
$19.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Acetal, Valved, 3/16" ID
Buy More Save More
EW-06360-12 Mfr # PMCD1603
COMPARE
3/16 Acetal 1/8
$17.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Acetal, Valved, 1/8" Flow Size, 1/4" ID
Buy More Save More
EW-06360-15 Mfr # PMCD1604
COMPARE
1/4 Acetal 1/8
$17.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 1/16" ID
Buy More Save More
EW-06360-19 Mfr # PMCD1701
COMPARE
1/16 Acetal 1/8
$13.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Body, Acetal, Valved, 1/8" Flow Size, 1/4" ID
Buy More Save More
EW-06360-25 Mfr # PMCD1704
COMPARE
1/4 Acetal 1/8
$12.30USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Acetal, Valved, 1/4" Flow Size, 1/4" ID
Buy More Save More
EW-06360-55 Mfr # PLCD16004
COMPARE
1/4 Acetal 1/4
$21.75USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Acetal, Valved, 5/16" ID
Buy More Save More
EW-06360-57 Mfr # PLCD16005
COMPARE
5/16 Acetal 1/4
$22.25USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 3/8" ID
Buy More Save More
EW-06360-70 Mfr # PLCD17006
COMPARE
3/8 Acetal 1/4
$11.60USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Body, Panel-Mount, Acetal, Straight-Through, 1/8" ID
Buy More Save More
EW-06361-04 Mfr # PMC1602
COMPARE
1/8 Acetal 1/8
$16.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Acetal, Straight-Through, 1/8" ID
Buy More Save More
EW-06361-11 Mfr # PMC1702
COMPARE
1/8 Acetal 1/8
$9.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Acetal, Straight-Through, 3/16" ID
Buy More Save More
EW-06361-12 Mfr # PMC1703
COMPARE
3/16 Acetal 1/8
$8.60USD / Each
CPC (Colder) Quick-Disconnect, Hose Barb Body, Panel-Mount, Acetal, Straight-Through, 1/4" Flow Size, 1/4" ID
Buy More Save More
EW-06361-46 Mfr # PLC16004
COMPARE
1/4 Acetal 1/4
$19.80USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Acetal, Straight-Through, 5/16" ID
Buy More Save More
EW-06361-52 Mfr # PLC17005
COMPARE
5/16 Acetal 1/4
$9.90USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Acetal, Straight-Through, 3/8" ID
Buy More Save More
EW-06361-53 Mfr # PLC17006
COMPARE
3/8 Acetal 1/4
$9.40USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Body, Panel-Mount, Polypropylene, Valved, 1/16" ID
Buy More Save More
EW-06364-09 Mfr # PMCD160112
COMPARE
1/16 Polypropylene 1/8
$19.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 1/8" ID
Buy More Save More
EW-06364-20 Mfr # PMCD170212
COMPARE
1/8 Polypropylene 1/8
$18.40USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Body, Panel-Mount, Polypropylene, Valved, 1/4" Flow Size, 1/4" ID
Buy More Save More
EW-06364-55 Mfr # PLCD1600412
COMPARE
1/4 Polypropylene 1/4
$23.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Body, Panel-Mount, Polypropylene, Valved, 3/8" ID
Buy More Save More
EW-06364-60 Mfr # PLCD1600612
COMPARE
3/8 Polypropylene 1/4
$24.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Body, Polypropylene, Valved, 1/4" Flow Size, 1/4" ID
Buy More Save More
EW-06364-65 Mfr # PLCD1700412
COMPARE
1/4 Polypropylene 1/4
$19.60USD / Each
Showing 1 - 20 of 38