CPC® Quick-Disconnect Fittings, Hose Barb Inserts
No more struggling—just press thumb latch
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
$3.50 - $385.00USD / Each
42 variations of this product are available.
See All Product Options Below
42 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Quick-Disconnect Flow Size (in)
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Valved, 1/16" ID
Buy More Save More
EW-06360-36 Mfr # PMCD2201
COMPARE
1/16 Acetal 1/8
$13.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Straight-Through, 1/16" ID
Buy More Save More
EW-06360-38 Mfr # PMC2201
COMPARE
1/16 Acetal 1/8
$5.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Valved, 1/8" ID
Buy More Save More
EW-06360-40 Mfr # PMCD2202
COMPARE
1/8 Acetal 1/8
$11.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Straight-Through, 1/8" ID
Buy More Save More
EW-06360-42 Mfr # PMC2202
COMPARE
1/8 Acetal 1/8
$3.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Straight-Through, 3/16" ID
Buy More Save More
EW-06360-44 Mfr # PMC2203
COMPARE
3/16 Acetal 1/8
$3.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Acetal, Valved, 1/8" Flow Size; 1/4" ID
Buy More Save More
EW-06360-45 Mfr # PMCD2204
COMPARE
1/4 Acetal 1/8
$10.60USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Acetal, Valved, 1/4" Flow Size; 1/4" ID
Buy More Save More
EW-06360-80 Mfr # PLCD22004
COMPARE
1/4 Acetal 1/4
$9.60USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Acetal, Straight-Through, 1/4" Flow Size, 1/4" ID
Buy More Save More
EW-06360-82 Mfr # PLC22004
COMPARE
1/4 Acetal 1/4
$3.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Straight-Through, 5/16" ID
Buy More Save More
EW-06360-84 Mfr # PLC22005
COMPARE
5/16 Acetal 1/4
$4.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Acetal, Straight-Through, 3/16" ID
Buy More Save More
EW-06361-23 Mfr # PMC4203
COMPARE
3/16 Acetal 1/8
$10.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Panel-Mount, Acetal, Valved, 1/8" ID
Buy More Save More
EW-06361-34 Mfr # PMCD4202
COMPARE
1/8 Acetal 1/8
$15.80USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Acetal, Straight-Through, 1/4" ID
Buy More Save More
EW-06361-58 Mfr # PLC42004
COMPARE
1/4 Acetal 1/4
$8.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Acetal, Valved, 1/4" ID
Buy More Save More
EW-06361-71 Mfr # PLCD42004
COMPARE
1/4 Acetal 1/4
$17.70USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Polypropylene, Valved, 3/8" ID
Buy More Save More
EW-06361-93 Mfr # PLCD4200612
COMPARE
3/8 Polypropylene 1/4
$20.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Polypropylene, Straight-Through, 1/16" ID
Buy More Save More
EW-06364-39 Mfr # PMC220112
COMPARE
1/16 Polypropylene 1/8
$7.30USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Straight-Through, 1/8" Flow Size, 1/4" ID
Buy More Save More
EW-06364-47 Mfr # PMC220412
COMPARE
1/4 Polypropylene 1/8
$6.80USD / Each
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Acetal, Straight-Through, 1/8" ID; 25/Pk EW-31307-14 Mfr # PMC4202
COMPARE
1/8 Acetal 1/8
$197.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Acetal, Straight-Through, 5/16" ID; 25/Pk EW-31307-49 Mfr # PLC42005
COMPARE
5/16 Acetal 1/4
$218.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Hosebarb Insert, Panel-Mount, Acetal, Valved, 3/8" ID; 25/Pk EW-31307-62 Mfr # PLCD42006
COMPARE
3/8 Acetal 1/4
$373.00USD / Pkg of 25
CPC (Colder) Quick-Disconnect Fitting, Hose Barb Insert, Panel-Mount, Polypropylene, Straight-Through, 3/8" ID; 25/Pk EW-31307-65 Mfr # PLC4200612
COMPARE
3/8 Polypropylene 1/4
$299.00USD / Pkg of 25
Showing 1 - 20 of 42