CPC® Quick-Disconnect Fittings, Threaded Bodies
No more struggling—just press thumb latch
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
$9.10 - $18.20USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Quick-Disconnect Flow Size (in) Thread size NPT(M) (port 1) (in)
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Acetal, Valved, 1/8" NPT(M)
Buy More Save More
EW-06359-70 Mfr # PMCD1002
COMPARE
Acetal 1/8 1/8
$12.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Thread Body, Acetal, Valved, 1/8" Flow Size, 1/4" NPT(M)
Buy More Save More
EW-06359-71 Mfr # PMCD1004
COMPARE
Acetal 1/8 1/4
$12.30USD / Each
CPC (Colder) Quick-Disconnect Fitting, Thread Body, Acetal, Valved, 1/4" Flow Size, 1/4" NPT(M)
Buy More Save More
EW-06359-72 Mfr # PLCD10004
COMPARE
Acetal 1/4 1/4
$11.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Acetal, Valved, 3/8" NPT(M)
Buy More Save More
EW-06359-73 Mfr # PLCD10006
COMPARE
Acetal 1/4 3/8
$11.00USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Polypropylene, Valved, 1/8" NPT(M)
Buy More Save More
EW-06359-80 Mfr # PMCD100212
COMPARE
Polypropylene 1/8 1/8
$13.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Thread Body, Polypropylene, Valved, 1/8" Flow Size, 1/4" NPT(M)
Buy More Save More
EW-06359-81 Mfr # PMCD100412
COMPARE
Polypropylene 1/8 1/4
$14.40USD / Each
CPC (Colder) Quick-Disconnect Fitting, Thread Body, Polypropylene, Valved, 1/4" Flow Size, 1/4" NPT(M)
Buy More Save More
EW-06359-82 Mfr # PLCD1000412
COMPARE
Polypropylene 1/4 1/4
$15.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Polypropylene, Valved, 3/8" NPT(M)
Buy More Save More
EW-06359-83 Mfr # PLCD1000612
COMPARE
Polypropylene 1/4 3/8
$18.20USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Acetal, Straight-Through, 1/8" NPT(M)
Buy More Save More
EW-06361-00 Mfr # PMC1002
COMPARE
Acetal 1/8 1/8
$9.10USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Acetal, Straight-Through, 1/8" Flow Size, 1/4" NPT(M)
Buy More Save More
EW-06361-01 Mfr # PMC1004
COMPARE
Acetal 1/8 1/4
$9.50USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Acetal, Straight-Through, 1/4" Flow Size, 1/4" NPT(M)
Buy More Save More
EW-06361-42 Mfr # PLC10004
COMPARE
Acetal 1/4 1/4
$14.30USD / Each
CPC (Colder) Quick-Disconnect Fitting, Threaded Body, Acetal, Straight-Through, 3/8" NPT(M)
Buy More Save More
EW-06361-43 Mfr # PLC10006
COMPARE
Acetal 1/4 3/8
$10.80USD / Each
Showing 1- 12 of 12