CPC® Quick-Disconnect Fittings with Automatic Shutoff Valve, Polysulfone
Eliminate the need for pinch clamps
  • Body and seal materials are made of USP Class VI-compliant and BSE/TSE-free materials
  • Valves restrict flow when fitting halves are disconnected
  • Fitting body constructed of durable polysulfone
These CPC® quick-disconnect fittings feature an automatic shutoff valve which closes off the flow path at disconnection, protecting valuable media while eliminating the need for pinch clamps. The thumb latch enables easy one-handed operation. An audible click is heard when the body and insert are coupled to confirm a secure, leak-free connection.
For a complete fitting, order both a body and insert (order separately).
$245.00 - $336.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hosebarb Body, Polysulfone, 1/4" ID; 10/Pk
Buy More Save More
EW-31313-01 Mfr # HFCD17439M
COMPARE
1/4 Polysulfone Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Body, Polysulfone, 1/4" ID; 10/Pk
$336.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hosebarb Body, Polysulfone, 3/8" ID; 10/Pk
Buy More Save More
EW-31313-02 Mfr # HFCD17639M
COMPARE
3/8 Polysulfone Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Body, Polysulfone, 3/8" ID; 10/Pk
$311.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hosebarb Body, Polysulfone, 1/2" ID; 10/Pk
Buy More Save More
EW-31313-03 Mfr # HFCD17839M
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Body, Polysulfone, 1/2" ID; 10/Pk
$334.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Insert, Polysulfone, 1/4" ID; 10/Pk
Buy More Save More
EW-31313-21 Mfr # HFCD22439M
COMPARE
1/4 Polysulfone Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Insert, Polysulfone, 1/4" ID; 10/Pk
$255.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Insert, Polysulfone, 3/8" ID; 10/Pk
Buy More Save More
EW-31313-22 Mfr # HFCD22639M
COMPARE
3/8 Polysulfone Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Insert, Polysulfone, 3/8" ID; 10/Pk
$245.00USD / Pkg of 10
CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Insert, Polysulfone, 1/2" ID; 10/Pk
Buy More Save More
EW-31313-23 Mfr # HFCD22839M
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Insert, Polysulfone, 1/2" ID; 10/Pk
$255.00USD / Pkg of 10