CPC® Sanitary Steam-in-Place Connectors, Two-Cycle
Ensure a sterile connection
  • One disposable connector for steam sterilization and microbial process
  • Reduces down time and cleaning expenses
As part of a single-use system, these CPC® sanitary Steam-in-Place disposable connectors enhance the sterility of your bioprocessing line. The innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections, and eliminating the need for a laminar flow hood. Terminations on these connectors are set to industry standards for bio-reactors and popular sizes of flexible tubing. All wetted surfaces meet requirements for USP Class VI, and physicochemical tests. All units are made of animal-free polysulfone resin.
These steam-in-place connectors offer "Steam On" and "Steam Off" functionality by allowing the valve to return to the steam position and enabling a second steam cycle after media transfer. The "steam off" disconnection of disposable systems minimizes the risk of cross-contamination caused by reusable system components.
The two-cycle steam-thru connectors offer the additional benefit of both “steam on” and “steam off” functionality. The valve can return to the steam position to allow a second steam cycle after media transfer.
$171.00 - $841.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 3) (in) Sanitary Clamp Size (Port 2) (in) Description
Availability Pricing
CPC (Colder) Sanitary Steam-in-Place Connector, Two-Cycle, 3/8" ID x 3/4" x 1-1/2" Sanitary; 5/Pk EW-31309-62 Mfr # STC2020200
COMPARE
3/8 1-1/2 Sanitary Steam-in-Place Connector, Two-Cycle, 3/8" ID x 3/4" x 1-1/2" Sanitary; 1/Ea
$841.00USD / Pkg of 5
CPC (Colder) Sanitary Steam-in-Place Connector, Two-Cycle, 1/2" ID x 3/4" x 3/4" Sanitary EW-31604-36 Mfr # STC2020000
COMPARE
1/2 3/4 Sanitary Steam-in-Place Connector, Two-Cycle, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Ea
$171.00USD / Each