CPS Portable Digital Vacuum Gauge
Provide constant indication and monitoring of the evacuation process from atmospheric down to 0 micron
  • VG100A
  • Range from atmospheric to 25 microns
  • Nine high-intensity LEDs indicate vacuum level
  • VG200
  • Range from atmospheric to 0 microns
  • Large 5-digit LCD
  • Readout in microns, in. Hg, or mBar
  • Continuous measurement indication
This advanced vacuum measurement instrument provide constant indication and monitoring of the evacuation process from atmospheric down to 0 micron. Advanced surface mount technology automatically compensates for ambient changes in the environment ensuring accuracy and consistency in performance. Rugged field-cleanable sensor is able to withstand 400 PSI of positive pressure. Instrument is CE approved.
$275.25USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display
Availability Pricing
CPS Products VG200 Portable Digital Vacuum Gauge, LCD, atmospheric to 0 microns EW-42500-08 Mfr # VG200
COMPARE
5-digit LCD
$275.25USD / Each