CPS Refrigerant Charging Hoses
1/4" Standard charging hoses
  • All-weather flexibility and strength
  • Extra deep knurling on nuts for easy grip
  • Available with MicroSeal™ or ball valve fittings
  • Leak-proof eight-sided crimp
  • Blow-off grooves channel refrigerant to prevent spray back
The hose has 1/4" SAE fittings with a 45° bend on one end. It can operate at 750 psi working pressure with a burst pressure of 3000 psi.
$39.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Standard Refrigerant Charge Hose, 3 ft/90 cm
$39.50USD / Each