Crosstex Sani-Roll Sterilization Tubing 64669
Convenient and economical way to package instruments
  • For use in steam, chemical vapor and EtO sterilizers
Sani-Rolls have easy to read color process indicators to signal exposure to sterilization conditions. Blue tinted plastic aids in the detection of puncture holes caused by pointed/sharp instruments. Complete paper and film separation and a triple seal.
$123.60 - $184.90USD / Case of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in)
Availability Pricing
Crosstex Sani-Roll Sterilization Tubing, Paper/Plastic, 2 Inx100 Ft EW-10703-01 Mfr # SCT2
COMPARE
1200 2
$149.90USD / Case of 1
Crosstex Sani-Roll Sterilization Tubing, Paper/Plastic, 3 In X 100 Ft EW-10703-02 Mfr # SCT3
COMPARE
1200 3
$123.60USD / Case of 1
Crosstex Sani-Roll Sterilization Tubing, Paper/Plastic, 4 In X 100 Ft EW-10703-03 Mfr # SCT4
COMPARE
1200 4
$144.50USD / Case of 1
Crosstex Sani-Roll Sterilization Tubing, Paper/Plastic, 6 In X100 Ft EW-10703-04 Mfr # SCT6
COMPARE
1200 5
$184.90USD / Case of 1