This product is now discontinued, View Related Products
Crosstex Steri-Tube Sterilization Tubing
Used in steam, chemical vapor, or dry heat up to 425°F ( 218°C)
Manufactured from clear, puncture-resistant 1.5 mil nylon. Easily sealed with heat sealer or process indicator tape (sold separately), Sani-Tubes are an economical and convenient way to package items for sterilization. Available with or without process indicators. Latex-free.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in)
Availability Pricing
COMPARE
1200 2
Call For Price
COMPARE
1200 3
Call For Price
COMPARE
1200 4
Call For Price
COMPARE
1200 5
Call For Price
COMPARE
1200 2
Call For Price
COMPARE
1200 3
Call For Price
COMPARE
1200 4
Call For Price
COMPARE
1200 5
Call For Price
Showing 1- 8 of 8