Crystal XP2i Digital Pressure Gauges
Accuracy not affected from 14 to 122°F (-10 to 50°C)
  • Start and stop multiple collection runs from the XP2i keypad
  • Replaces five or more digital test gauges
  • Marine grade enclosure resists corrosion
  • IP67 certified
The XP2i digital pressure gauges provide an accurate reading of 0.1% from 14 to 122°F (-10 to 50°C). The gauges are easy to operate with the use of three simple function buttons. A fast pressure safety valve (PSV) mode allows you to capture and display the relief pressure even if you are not looking at the gauge when the valve opens.
Sealed against dust and water, the gauges are IP67 certified for 30 minutes of full submersion under 3 ft (1 m) of water. They are designed to survive falls with no affect on accuracy and feature a corrosion resistant nickel-plated housing. A large, high contrast LCD with backlight makes it easy to read the display anywhere.
$1,149.00 - $1,999.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Crystal 10KPSIXP2I Digital Pressure Gauge, 10,000 psi EW-68436-69 Mfr # 10KPSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 10000
$1,213.00USD / Each
Crystal 15PSIXP2I Digital Pressure Gauge, 15 psi EW-68436-70 Mfr # 15PSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 15
$1,199.00USD / Each
Crystal 1KPSIXP2I Digital Pressure Gauge, 1000 psi EW-68436-71 Mfr # 1KPSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 1000
$1,149.00USD / Each
Crystal 100PSIXP2I Digital Pressure Gauge, 100 psi EW-68436-81 Mfr # 100PSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 100
$1,149.00USD / Each
Crystal 2KPSIXP2I Digital Pressure Gauge, 2000 psi EW-68436-82 Mfr # 2KPSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 2000
$1,999.00USD / Each
Crystal 300PSIXP2I Digital Pressure Gauge, 300 psi EW-68436-83 Mfr # 300PSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 300
$1,149.00USD / Each
Crystal 30PSIXP2I Digital Pressure Gauge, 30 psi EW-68436-84 Mfr # 30PSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 30
$1,215.00USD / Each
Crystal 3KPSIXP2I Digital Pressure Gauge, 3000 psi EW-68436-85 Mfr # 3KPSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 3000
$1,184.00USD / Each
Crystal 500PSIXP2I Digital Pressure Gauge, 500 psi EW-68436-86 Mfr # 500PSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 500
$1,199.00USD / Each
Crystal 5KPSIXP2I Digital Pressure Gauge, 5000 psi EW-68436-87 Mfr # 5KPSIXP2I
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 5000
$1,243.20USD / Each
Showing 1- 10 of 10