This product is now discontinued, View Related Products

Danger: Combustible Signs

Protect your workers by clearly displaying possible combustible hazards
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MCHL141VS Safety Sign, Danger - Combustible, 7" x 10", Adhesive Vinyl EW-40711-87 Mfr # MCHL141VS
COMPARE
7 Vinyl 10
Call For Price
Accuform MCHL141VP Safety Sign, Danger - Combustible, 7" x 10", Plastic EW-40711-88 Mfr # MCHL141VP
COMPARE
7 Plastic 10
Call For Price
Accuform MCHL141VA Safety Sign, Danger - Combustible, 7" x 10", Aluminum EW-40711-89 Mfr # MCHL141VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MCHL103VS Safety Sign, Danger - Combustible, 10" x 14", Adhesive Vinyl EW-40711-90 Mfr # MCHL103VS
COMPARE
10 Vinyl 14
Call For Price
Accuform MCHL103VP Safety Sign, Danger - Combustible, 10" x 14", Plastic EW-40711-91 Mfr # MCHL103VP
COMPARE
10 Plastic 14
Call For Price
Accuform MCHL103VA Safety Sign, Danger - Combustible, 10" x 14", Aluminum EW-40711-92 Mfr # MCHL103VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat