This product is now discontinued, View Related Products

Danger: Stay Clear Signs

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MEQM190VS Safety Sign, Danger - Stay Clear, 7" X 10", Adhesive Vinyl EW-40710-96 Mfr # MEQM190VS
COMPARE
7 Adhesive Vinyl 10
Call For Price
Accuform MEQM190VP Safety Sign, Danger - Stay Clear, 7" X 10", Plastic EW-40710-97 Mfr # MEQM190VP
COMPARE
7 Plastic 10
Call For Price
Accuform MEQM190VA Safety Sign, Danger - Stay Clear, 7" X 10", Aluminum EW-40710-98 Mfr # MEQM190VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MEQM078VS Safety Sign, Danger - Stay Clear, 10" X 14", Adhesive Vinyl EW-41014-96 Mfr # MEQM078VS
COMPARE
10 Adhesive Vinyl 14
Call For Price
Accuform MEQM078VP Safety Sign, Danger - Stay Clear, 10" X 14", Plastic EW-41014-97 Mfr # MEQM078VP
COMPARE
10 Plastic 14
Call For Price
Accuform MEQM078VA Safety Sign, Danger - Stay Clear, 10" X 14", Aluminum EW-41014-98 Mfr # MEQM078VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat