DC-Powered Mini Diaphragm Pumps
DC-powered pumps with PVDF pump head
  • High resolution with a flow range of 0.055 to 38 mL/min
  • Choose between 12 VDC or 24 VDC power—pulse, start/stop, or proportional control
  • Connect 1/8" x 1/4" tubing directly to pump ports
Choose a pump with one of the following control methods:
Pulse: 1 stroke per pulse—0 to 720 strokes per minute control capability
4 to 20 mA: proportional control based on mA signal
1 to 5 VDC: proportional control based upon a 1 to 5 VDC input signal
Start/stop: pump stops during contact input
$236.40 - $243.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VDC) Description
Availability Pricing
COMPARE
12 HRP54V-1P3 DC-Powered Diaphragm Pump; 12 VDC, Pulse
$243.50USD / Each
COMPARE
24 HRP54V-2P3 DC-Powered Diaphragm Pump; 24 VDC, Pulse
$243.50USD / Each
COMPARE
12 HRP54V-1A3 DC-Powered Diaphragm Pump; 12 VDC, 4-20 mA Proportional
$236.40USD / Each
COMPARE
24 HRP54V-2A3 DC-Powered Diaphragm Pump; 24 VDC, 4-20 mA Proportional
$236.40USD / Each
COMPARE
12 HRP54V-1V3 DC-Powered Diaphragm Pump; 12 VDC, 0-5 V Proportional
$236.40USD / Each
COMPARE
24 HRP54V-2V3 DC-Powered Diaphragm Pump; 24 VDC, 0-5 V Proportional
$243.50USD / Each
COMPARE
12 HRP54V-1S3 DC-Powered Diaphragm Pump; 12 VDC, Start/Stop
$236.40USD / Each
COMPARE
24 HRP54V-2S3 DC-Powered Diaphragm Pump; 24 VDC, Start/Stop
$243.50USD / Each
Showing 1- 8 of 8