Dedicated Transit-Time Flowmeter Kits for Liquids
Extremely simple installation and set-up
  • Economical non-intrusive flow measurement
  • Suitable for a wide range of pipe sizes and materials
  • Velocity, volumetric, and totalized flow outputs
The Panemetrics AquaTrans AT600 liquid flow ultrasonic transmitter is designed to be installed right at the process measurement point. Designed specifically for water and wastewater applications in full pipes, the flowmeter has no moving parts and requires minimal maintenance. Onboard microprocessor uses technology for long-term, drift-free operation. Automatic adjustment to changing fluid properties and dynamically configured operating software simplify programming.
The clamp-on transducers are easily installed, and combined with the electronics, provide better than 1% of reading accuracy in most applications. Choose from three different kits most suitable for your pipe, covering sizes from 2" to 24" (60 to 600 mm).
Flowmeter Fluid Types: Conductive fluids, including most clean liquids, and many liquids with small amounts of entrained solids or gas bubbles.
Water and wastewater applications include potable water, sewage, discharge water, cooling and heating water, irrigation, and treated water.
$2,676.50 - $3,246.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Communications Output Pipe Size
Availability Pricing
GE Panametrics AT6 Transit-Time Ultrasonic Flowmeter, for Pipes 8" to 24" EW-32499-70 Mfr # AT6-C1-CR05-24IN-2-1AAT01E-0
COMPARE
4 to 20 mA, alarm contact, and totalizer output
$3,246.50USD / Each
GE Panametrics AT6 Transit-Time Ultrasonic Flowmeter, for Pipes 4" to 12" EW-32499-71 Mfr # AT6-C1-CR10-12IN-2-1AAT01E-0
COMPARE
4 to 20 mA, alarm contact, and totalizer output
$3,226.50USD / Each
GE Panametrics AT6 Transit-Time Ultrasonic Flowmeter, for Pipes 2" to 6" EW-32499-72 Mfr # AT6-C1-AT20-6IN-2-1AAT01E-0
COMPARE
4 to 20 mA, alarm contact, and totalizer output
$2,676.50USD / Each