This product is now discontinued, View Related Products
Defelsko Coating Thickness Gauge - DF50201 and DF50202
DF50201
For hot dip galvanizing, hard chrome, metalizing, paint, enamel, plastic coating, on steel
DF50202
For electroplating, thin paint films, phosphating, on steel
FEATURES
  • Positive visual and audible indicators to designate when thickness reading is established
  • "V" grooves in probe housing and instrument base to allow correct positioning on cylindrical objects
  • Go-No Go button can be preset for rapid measurement
  • New overall design, full supported, positive positioning, no pivoting tendencies during measurement
  • Refined magnetic and balanced arm assemblies, improved resolution and accuracy
  • Extra rugged housing, not affected by mechanical shock, water, acid or solvents
  • Probe contact and dial rotation all in one finger operation
  • Explosion proof, one year warranty
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Scale