This product is now discontinued, View Related Products

DESCO Statshield ESD Smocks

Conductive knitted cuff eliminates the need for wristband
  • Hip-to-cuff grounding design provides hands-free grounding
  • Constructed of 9% carbon conductive nylon fibers for maximum charge dissipation
These smocks allow coil cords to snap to the hip of the smock enabling operators to be grounded hands-free. Smocks feature panel-to-panel conductivity preventing an isolated charged conductor in ESD-protected areas. Each smock has two lower pockets and one left breast pocket, and can withstand over one hundred washings without losing its static resistance.

Compliance: ANSI/ESD S20.20 - 2007; ESD STM 2.1
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Color Length (in) Size
Availability Pricing
Desco 73750 Static Control Smock, Small, 30" L EW-26810-52 Mfr # 73750
COMPARE
Blue 30 Small
Call For Price
Desco 73755 Static Contol Smock, Medium, 30" L EW-26810-53 Mfr # 73755
COMPARE
Blue 30 Medium
Call For Price
Desco 73760 Static Control Smock, Large, 30" L EW-26810-54 Mfr # 73760
COMPARE
Blue 30 Large
Call For Price
Desco 73765 Static Control Smock, X-Large, 30" L EW-26810-55 Mfr # 73765
COMPARE
Blue 30 X-Large
Call For Price
Desco 73611 Static Control Smock, Small, 42" L EW-26810-56 Mfr # 73611
COMPARE
Blue 42 Small
Call For Price
Desco 73612 Static Control Smock, Medium, 42" L EW-26810-57 Mfr # 73612
COMPARE
Blue 42 Medium
Call For Price
Desco 73613 Static Control Smock, Large, 42" L EW-26810-58 Mfr # 73613
COMPARE
Blue 42 Large
Call For Price
Desco 73614 Static Control Smock, X-Large, 42" L EW-26810-59 Mfr # 73614
COMPARE
Blue 42 X-Large
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat