Diba Click-N-Seal® Adapters with M6 ISO(M) Thread for Softwall Tubing
Hand tighten fittings without tools to create a secure seal
  • When the cap clicks, you know the seal is tight
These unique self-torquing fittings create a secure thread to barb connection without the use of any tools. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. Select from polycarbonate (PC) or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
$64.90 - $130.00USD / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Color Fitting Material
Availability Pricing
Diba Click-N-Seal™ Adapter, M6 ISO(M) to 1/16" ID, black PC; 10/pk EW-21941-23 Mfr # 008NB16-CS6B
COMPARE
1/16 Black Polycarbonate
$64.90USD / Pkg of 10
Diba Click-N-Seal™ Adapter, M6 ISO(M) to 1/16" ID, white PC; 10/pk EW-21941-24 Mfr # 008NB16-CS6H
COMPARE
1/16 White Polycarbonate
$64.90USD / Pkg of 10
Diba Click-N-Seal™ Adapter, M6 ISO(M) to 1/16" ID, red PC; 10/pk EW-21941-25 Mfr # 008NB16-CS6R
COMPARE
1/16 Red Polycarbonate
$64.90USD / Pkg of 10
Diba Click-N-Seal™ Ultra Adapter, M6 ISO(M) to 1/16" ID, natural PEEK™; 10/pk EW-21941-27 Mfr # 008NB16-KT6L
COMPARE
1/16 Natural PEEK
$130.00USD / Pkg of 10
Diba Click-N-Seal™ Adapter, M6 ISO(M) to 1/8" ID, black PC; 10/pk EW-21941-45 Mfr # 008NB32-CS6B
COMPARE
1/8 Black Polycarbonate
$67.00USD / Pkg of 10
Diba Click-N-Seal™ Adapter, M6 ISO(M) to 1/8" ID, white PC; 10/pk EW-21941-46 Mfr # 008NB32-CS6H
COMPARE
1/8 White Polycarbonate
$67.00USD / Pkg of 10
Diba Click-N-Seal™ Adapter, M6 ISO(M) to 1/8" ID, red PC; 10/pk EW-21941-47 Mfr # 008NB32-CS6R
COMPARE
1/8 Red Polycarbonate
$67.00USD / Pkg of 10
Diba Click-N-Seal™ Ultra Adapter, M6 ISO(M) to 1/8" ID, natural PEEK™; 10/pk EW-21941-51 Mfr # 008NB32-KT6L
COMPARE
1/8 Natural PEEK
$130.00USD / Pkg of 10
Showing 1- 8 of 8