Dickson 4 inch Compact Pressure Chart Recorders
4" Compact Pressure Recorders Weather-resistant, NEMA 4X-rated enclosure withstands harsh outdoor environments
  • Compact 4" circular chart records pressure up to 500 psi
  • Choose from 24-hour or 7-day recording times
  • Battery operated for easy portability
Place these compact, portable pressure recorders anywhere a written record of pressure is needed. The durable Velox PBT NEMA 4X-rated case protects against corrosion, windblown dust and rain, splashing water, and hose-directed water. Recorders are ideal for investigating and monitoring systems for gas, steam, and water leaks; documenting system pressure at water treatment/ purification facilities; and measuring flow of water wells. Unique door closure ensures weather resistance.
Recorder operates for up to one year on a single AA battery. On/off switch provides battery savings. Battery holder is located inside the door for easy battery changes.
Note: These pressure recorders are not suitable for monitoring
ammonia, acids, or other corrosive substances.
$642.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chart Diameter (in) Chart Speed Max Pressure (PSI)
Availability Pricing
Dickson PW470 Pressure Chart Recorder, 0 to 100 PSI EW-49010-00 Mfr # PW470
COMPARE
4 24 hours or 7 days (switchable) 100
$642.00USD / Each
Dickson PW474 Pressure Chart Recorder, 0 to 200 PSI
Always in Stock
EW-49010-01 Mfr # PW474
COMPARE
4 24 hours or 7 days (switchable) 200
$642.00USD / Each
Dickson PW476 Pressure Chart Recorder, 0 to 300 PSI
Always in Stock
EW-49010-02 Mfr # PW476
COMPARE
4 24 hours or 7 days (switchable) 300
$642.00USD / Each
Dickson PW479 Pressure Chart Recorder, 0 to 500 PSI EW-49010-03 Mfr # PW479
COMPARE
4 24 hours or 7 days (switchable) 500
$642.00USD / Each