Digi-Sense Adjustable Angle Dual-Scale Industrial Bimetal Thermometers
Cole-Parmer Process grade thermometers are silicone-free
  • Greater temperature range
  • Anti-parallax dial for accurate readings
  • Hermetically sealed per ASME B40.3 standard to prevent fogging and protect against tough environments
  • Rugged 304 stainless steel construction with a clear glass lens
  • Recalibrate using the external reset screw
Accuracy is ±1% full-scale when stem is immersed 2" in liquids or 4" in gases (immerse models with a 2.5" stem length 2" in gases). Measured stem length from the top of the threads to the bottom tip of the stem. Stem dia is 0.25"; threaded connection is 1/2" NPT(M). All thermometers meet or exceed ASME B40.3 standards. Maximum environmental temperature for glass lens thermometers is 500°F (148°C).
Discounts: Save 10% on 10-24 assorted thermometers, 15% on 25 to 49, and 20% on 50 or more.
$91.00 - $151.00USD / Each
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
3 300 50
$101.00USD / Each
COMPARE
3 550 50
$101.00USD / Each
COMPARE
3 250 0
$101.00USD / Each
COMPARE
3 550 50
$101.00USD / Each
COMPARE
3 250 0
$101.00USD / Each
COMPARE
3 125 25
$101.00USD / Each
COMPARE
3 300 50
$101.00USD / Each
COMPARE
3 550 50
$101.00USD / Each
COMPARE
3 250 0
$99.50USD / Each
COMPARE
3 125 25
$108.00USD / Each
COMPARE
3 550 50
$111.00USD / Each
COMPARE
3 250 0
$128.00USD / Each
COMPARE
3 300 50
$128.00USD / Each
COMPARE
3 550 50
$128.00USD / Each
COMPARE
5 125 25
$118.00USD / Each
COMPARE
5 550 50
$115.00USD / Each
COMPARE
5 250 0
$115.00USD / Each
COMPARE
5 250 0
$130.00USD / Each
COMPARE
5 550 50
$131.00USD / Each
COMPARE
5 250 0
$151.00USD / Each
Showing 1 - 20 of 31